Learn More
Medchemexpress LLC FAPI-4 | 2374782-02-0 | 99.5% | 872.91 | 10 MG
SDP
Supplier:  Medchemexpress LLC 10MGHY128643
Encompass_Preferred
FAPI-4 is a potent inhibitor of fibroblast activation protein (FAP), primarily used for targeting FAP in cancer research. Its application in 68Ga-FAPI-4 PET/CT provides a promising diagnostic method for imaging various cancers, demonstrating good tumor-to-background contrast ratios.
  • Potent fibroblast activation protein (FAP) inhibitor
  • Useful in cancer research
  • Promising diagnostic method for imaging various cancers with good tumor-to-background contrast ratios
  • For research use only
Catalog No. 50-003-65149