Learn More
Apexbio Technology LLC FAPI-74 2374782-76-8 25mg
SDP
Supplier:  Apexbio Technology LLC BA596925
Encompass_Preferred
FAPI-74 (CAS 2374782-76-8) is a small molecule positron emission tomography (PET) tracer that selectively targets fibroblast activation protein (FAP) a serine protease overexpressed in cancer-associated fibroblasts within the tumor microenvironment By binding to FAP FAPI-74 enables noninvasive imaging of FAP-expressing tissues facilitating tumor detection and characterization This compound is utilized in preclinical and clinical research to investigate tumor biology monitor disease progression and assess the potential of FAP as a diagnostic and therapeutic target in oncology studies
Catalog No. 50-000-88791