Learn More
Medchemexpress LLC FAPI-74 | 2374782-76-8 | 99.82% | 735.83 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY14717310MM/1ML
Encompass_Preferred
FAPI-74 is a PET (positron emission tomography) tracer targeting the fibroblast activation protein (FAP), primarily for research into FAP-positive tumors. It has been utilized in in vitro chelation with 18F-AlF and 68Ga, and in PET/CT imaging to aid tumor volume delineation for guiding radiotherapy in lung cancer within PANC-1 xenograft mice. The compound appears as an off-white to light yellow solid.
  • Targets fibroblast activation protein
  • Useful for FAP-positive tumor research
  • Can be used for in vitro chelation with 18F-AlF and 68Ga
  • Applied in PET/CT imaging to delineate tumor volume
  • Soluble in DMSO at 125 mg/mL (169.88 mM)
Catalog No. 50-003-59598