Learn More
Enzo Life Sciences Farnesylthioacetic acid (5mg). CAS: 135784-48-4
SDP
Supplier:  Enzo Life Sciences BMLG2220005
Encompass_Preferred
FTA potently and specifically inhibits methyl esterification of farnesylated proteins such as the γ subunit of transducin (70% at 10µM). It does not act as a substrate for methyl transferase. Alternative name: FTA. Purity: ≥98% (TLC). Solubility: Soluble in DMSO (>25mg/ml) or 100% ethanol (>25mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-0855