Learn More
Cayman Chemical Farnsyl Alcohol AzIde 100ug
SDP
Supplier:  Cayman Chemical 13269100 UG
Encompass_Preferred
Acts as a replacement for endogenously-produced farnesyl alcohol and becomes attached to proteins through normal biological processes, in cells or animals; the terminal azide group can be used to readily tag farnesylated proteins via click chemistry for subsequent analysis
Catalog No. 50-295-2918