Learn More
Cayman Chemical Farnsyl ThIosalcylc ACD A 5mg
SDP
Supplier:  Cayman Chemical 131755 MG
Encompass_Preferred
An inhibitor of Ras-mediated signaling that inhibits the growth of PANC-1 and U87 tumor cells with IC50 values of 20 and 10 µM, respectively
Catalog No. 50-295-1736