Learn More
Medchemexpress LLC Faropenem sodium | 122547-49-3 | 98.9% | C12H14NNaO5S | 1 ML
Supplier: Medchemexpress LLC HY7626010MM/1ML
Faropenem sodium is an orally bioavailable penem antibiotic that regulates inorganic phosphate transporter Npt1. It inhibits M. tuberculosis (MIC of 1.3 μg/mL) and B. anthracis.
- Orally bioavailable penem antibiotic
- Regulates inorganic phosphate transporter Npt1
- Inhibits M. tuberculosis (MIC of 1.3 μg/mL)
- Inhibits B. anthracis