Learn More
Cayman Chemical FaropenmsodIM salt hydrat 10mg
SDP
Supplier:  Cayman Chemical 3450710 MG
Encompass_Preferred
A penem antibiotic; active against a panel of 12 Gram-positive (MIC50s = 0.008-1 mg/L) and 15 Gram-negative (MIC50s = 0.06-16 mg/L) bacteria; reduces the number of lung CFUs in a mouse model of H. influenzae-induced respiratory tract infection
Catalog No. 50-295-0806