Learn More
Medchemexpress LLC HY-N0344 5mg Medchemexpress, Farrerol CAS:24211-30-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN03445MG
Encompass_Preferred
Medchemexpress, HY-N0344 5mg Farrerol CAS:24211-30-1 Farrerol is a bioactive constituent of Rhododendron, with broad activities such as anti-oxidative, anti-inflammatory, anti-tumor, neuroprotective and hepatoprotective effects[2][3][4][5][6]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-1332