Learn More
Medchemexpress LLC Farudodstat | 1035688-66-4 | 99.7% | 50 MG
SDP
Supplier:  Medchemexpress LLC HY12923950MG
Encompass_Preferred
Farudodstat (ASLAN003) is an orally active and potent Dihydroorotate Dehydrogenase (DHODH) inhibitor with an IC50 of 35 nM for the human DHODH enzyme. It inhibits protein synthesis by activating AP-1 transcription factors. Farudodstat induces apoptosis and significantly prolongs survival in acute myeloid leukemia (AML) xenograft mice.
  • Orally active and potent dihydroorotate dehydrogenase (DHODH) inhibitor
  • Inhibits protein synthesis via activation of AP-1 transcription factors
  • Induces apoptosis
  • Substantially prolongs survival in acute myeloid leukemia (AML) xenograft mice
Catalog No. 50-003-38851