Learn More
Abcam Fascaplysin, ATP-competitve CDK4 inhibitor, 1MG
Supplier:  Abcam AB1461771MG
Encompass_Preferred
MW 306.75 g/mol. Potent, selective ATP-competitive CDK4 inhibitor (IC₅₀ = 350 nM). Suppresses VEGF expression and inhibits endothelial cell proliferation. Induces apoptosis and cytotoxic autophagy in vitro.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4882