Learn More
MSC Weldas STEERSOtuff Leather Bib Apron, Side Split Cowhide, 36 in. x 42 in., 1 ea.
Supplier: MSC 35768985
This leather bib apron is designed to provide comprehensive heat and flame protection for the breast, belly, and legs during welding operations. Crafted from durable and pliable side split cowhide, it features an ergonomic design to ensure comfort while maintaining high resistance to heat and abrasion.
- Constructed from durable side split cowhide
- Heat and flame resistant material
- Double-locked stitched seams with high-strength 5-ply Kevlar® thread
- Reinforced stress points using metal rivets or double leather
- Coated rivets and snaps for electrical insulation
- Adjustable belt and quick-release strap system
- Measures 36 in. x 42 in.