Learn More
Medchemexpress LLC Fasiglifam | 1000413-72-8 | MFCD18251445 | 99.0% | 524.63 | C29H32O7S | 25 MG
SDP
Supplier:  Medchemexpress LLC HY1048025MG
Encompass_Preferred
Fasiglifam is a potent, selective, orally bioavailable agonist of the GPR40 (FFAR1) receptor used in research to enhance glucose-stimulated insulin secretion and probe metabolic signaling pathways. The compound is provided as a high-purity solid for laboratory use with defined storage recommendations.
  • Potent GPR40 agonist (EC50 72 nM).
  • Selective activity toward FFAR1, useful for receptor pharmacology studies.
  • Orally bioavailable compound appropriate for in vivo pharmacology.
  • High purity (98.97%), suitable for analytical and biological assays.
  • Supplied as a solid powder with recommended cold storage.
  • For research use only; not for human administration.
Catalog No. 50-002-44042