Learn More
Medchemexpress LLC FASN-IN-3 | 2097262-60-5 | 384.47 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYU00436100MG
Encompass_Preferred
FASN-IN-3 is a fatty acid synthase (FASN) inhibitor, extracted from patent US20170119786A1, compound 242A. It is intended for research use only.
  • Reduces IL-1 beta secretion from human PBMC stimulated with LPS or LTA.
  • Reduces IL-1 beta from LTA-stimulated monocytes.
  • Shows slight reduction of IL-1 beta after LPS stimulation.
  • Targets fatty acid synthase (FASN).
Catalog No. 50-004-45701