Learn More
Medchemexpress LLC HY-U00436 5mg Medchemexpress, FASN-IN-3 CAS:2097262-60-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYU004365MG
Encompass_Preferred
Medchemexpress, HY-U00436 5mg FASN-IN-3 CAS:2097262-60-5 FASN-IN-3 is a fatty acid synthase (FASN) inhibitor extracted from patent US20170119786A1, compound 242A. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3195