Learn More
Medchemexpress LLC FASN-IN-4 tosylate | 2095432-57-6 | 98.55% | 1 ML
SDP
Supplier:  Medchemexpress LLC HY12648A10MM/1ML
Encompass_Preferred
FASN-IN-4 tosylate is a potent inhibitor of fatty acid synthase (FASN) with an IC50 of 10 nM. It also inhibits SARS-CoV-2 with an EC50 of 18.6 nM, suggesting its potential for COVID-19 research through its ability to inhibit SARS-CoV-2 replication.
  • Potent FASN inhibitor
  • Inhibits SARS-CoV-2 replication
  • Potential for COVID-19 research
  • IC50 of 10 nM (FASN)
  • EC50 of 18.6 nM (SARS-CoV-2)
Catalog No. 50-003-41653