Learn More
Medchemexpress LLC HY-12648A 2mg Medchemexpress, FASN-IN-4 (tosylate) CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY12648A2MG
Encompass_Preferred
Medchemexpress, HY-12648A 2mg FASN-IN-4 (tosylate) CAS: FASN-IN-4 tosylate is a potent inhibitor of fatty acid synthase (FASN) extracted from patent WO 2012064642 A1, compound 29; has an IC50 of 10 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1109