Learn More
Nova Biomedical Corporation NOVA CDV Sample Cup, 100 µL Capacity, Compatible with BioProfile FAST CDV, 500 Cups
Supplier:  Nova Biomedical Corporation 65269
Encompass
The NOVA BIOMEDICAL CORPORATION FAST CDV Sample Cup is designed for accurate cell density and viability analysis, compatible with the Nova Biomedical BioProfile FAST CDV analyzer. With a 100 µL capacity, it prevents cell adherence and enhances mixing, ensuring reliable results for high-density cultures. The proprietary design optimizes sample integrity and allows waste containment for up to 500 cups, setting it apart from standard sample cups.
Catalog No. NC3592590