Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences Fast Green FCF, Certified, C.I. DcGf-12 5 GR
SDP

Supplier:  Electron Microscopy Sciences 15500

Encompass_Preferred

(Food Green 3)
CAS #2353-45-9 C₃₇H₃₄O₁₀N₂S₃Na₂ F.W. 808.86
Solubility: 4.0% Water; 9.0% Alcohol; 8.0% Cellosolve; 6.25% Glycol; 0.0% Xylene

The main advantage of this dye is its resistance to fading. Mammalian tissue stain for visualizing collagen, muscle, cystoplasma, cells and cornified epithelium.

Kornhauser, Stain Tech., 18,95 (1943); 20, 23 (1945);
Lillie, Arch. Path., 29, 705 (1940). Modified Feulgen stain for visualizing chromosomes in root tips and other meristematic tissue.
DeTomasi, Stain Tech., 11, 137 (1936). Used in plant histology.
O. Conn, "Biological Stains" 9th ed., Williams & Wilkens Co., Baltimore, 1977, pp. 252-3; Haynes, Stain Tech., 3, 40 (1928).

Catalog No. 50-980-470


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.