Learn More
Bioworld Fast Terrestrial Orchid Medium, 25 L
Supplier:  Bioworld 306301843
Encompass_Preferred
Fast Terrestrial Orchid MediumContains Macronutrients, Micronutrients, Vitamins, Sucrose, Fructose, Agar, and Supplements required to culture terrestrial orchidsCytokines and Auxins should be added as needed Provided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture testedWorking Concentration: 27.84 g/LpH: 5.2-6.0Partially soluble in water
Catalog No. 50-254-956