Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fasudil hydrochloride | 105628-07-7 | MFCD00943198 | 100.0% | 327.83 | C14H18ClN3O2S | 5 G
SDP

Supplier:  Medchemexpress LLC HY103415G

Encompass_Preferred

Fasudil hydrochloride is the monohydrochloride salt of fasudil, a small-molecule RhoA/ROCK inhibitor used in biochemical and cellular research to modulate kinase signaling. It is supplied as a solid powder with high stated purity and is suitable for in vitro assays and biochemical studies.

  • RhoA/ROCK inhibitor used in kinase and cellular assays
  • Monohydrochloride salt, solid powder
  • High stated purity (99.97%) for analytical and research use
  • Molecular formula: C14H18ClN3O2S; molecular weight: 327.83 g/mol
  • Suitable for biochemical assays and cell-based studies
  • Available in laboratory-scale pack sizes, including 5 g

Catalog No. 50-002-44909


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.