Learn More
Medchemexpress LLC Fasudil (hydrochloride) | 105628-07-7 | MFCD00943198 | 100.0% | 327.83 g·mol⁻¹ | C14H18ClN3O2S | 1 ML

Supplier: Medchemexpress LLC HY1034110MM 1ML
Fasudil hydrochloride is a small-molecule Rho-associated kinase (ROCK) inhibitor used in biochemical and cellular research. It inhibits ROCK1/2 and several protein kinases, and acts as a calcium channel antagonist and vasodilator. The compound is supplied as a purified solid or as a ready-to-use 10 mM solution in DMSO (1 mL). Molecular formula C14H18ClN3O2S; molecular weight 327.83 g·mol⁻¹. For research use only - not for human or clinical use.
- Ready-to-use 10 mM solution in DMSO, 1 mL.
- Also available as purified solid in multiple pack sizes.
- High purity (≈99.97%).
- Stable when stored sealed at 4°C; in solution store at -20°C to -80°C.
- Intended for biochemical and cellular assays targeting ROCK signaling.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.