Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fasudil hydrochloride | 105628-07-7 | MFCD00943198 | 100.0% | 327.83 g/mol | C14H18ClN3O2S | 100 MG
SDP

Supplier:  Medchemexpress LLC HY10341100MG

Encompass_Preferred

Fasudil hydrochloride is the hydrochloride salt of fasudil, a nonspecific RhoA/ROCK inhibitor used in research to investigate Rho-kinase-mediated signaling and kinase activity. It has CAS 105628-07-7, molecular formula C14H18ClN3O2S, and a molecular weight of 327.83 g/mol. The compound is supplied as a high-purity research reagent and is soluble in water and DMSO at the concentrations indicated on the product datasheet.

  • Nonspecific RhoA/ROCK inhibitor for biochemical and cellular studies.
  • Hydrochloride salt form for improved aqueous solubility.
  • High purity (99.97%) suitable for research applications.
  • Molecular weight 327.83 g/mol; formula C14H18ClN3O2S.
  • Solubility: H2O 55 mg/mL (requires ultrasonic), DMSO ≥ 31 mg/mL.
  • Intended for research use only; not for human or clinical use.

Catalog No. 50-002-46666


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.