Learn More
Selleck Chemical LLC Fatostatin S9785-10MM/1ML
SDP
Supplier:  Selleck Chemical LLC S978510MM/1ML
Encompass_Preferred
Fatostatin (125B11) a diarylthiazole derivative is a specific inhibitor of Sterol regulatory element binding proteins (SREBPs) activation Fatostatin binds to SCAP (SREBP cleavage-activating protein) and inhibits the ER-Golgi translocation of SREBPs Fatostatin suppresses growth and enhances apoptosis in cancer cells
Catalog No. 50-000-24585