Learn More
Selleck Chemical LLC Fatostatin HBr
SDP
Supplier:  Selleck Chemical LLC S8284200MG
Encompass_Preferred
Fatostatin HBr is an inhibitor of sterol regulatory element binding protein (SREBP) It impairs the activation of SREBP-1 and SREBP-2
Catalog No. 50-000-00566