Learn More
Cayman Chemical FatostatInhydrobromIde 5mg
SDP
Supplier:  Cayman Chemical 135625 MG
Encompass_Preferred
An inhibitor of SREBP activation, preventing SCAP-mediated escort of either SREBP-1 or SREBP-2 to the Golgi (IC50 = 5.6 µM); blocks SREBP-mediated gene expression; inhibits high glucose-induced upregulation of TGF-β in primary rat mesangial cells; reduces hepatic fat accumulation in ob/ob mice
Catalog No. 50-295-3612