Learn More
Medchemexpress LLC Fatp1-in-2 | 2650944-83-3 | 98.8% | 353.39 g/mol | C19H20FN5O | 5 MG

Supplier: Medchemexpress LLC HY1417005MG
This arylpiperazine-derived small molecule is a potent, orally active inhibitor of fatty acid transport protein 1 (FATP1), developed for use in biochemical and cellular studies of fatty acid transport and metabolic pathways. It demonstrates submicromolar activity in human and mouse assays and is supplied with high reported purity.
- Orally active fatty acid transport protein 1 (FATP1) inhibitor.
- Submicromolar potency: human IC50 = 0.43 μM, mouse IC50 = 0.39 μM.
- High reported purity suitable for research use (98.8% reported).
- Available in small-mass solid formats and DMSO solution formats for assay convenience.
- Molecular formula C19H20FN5O; molecular weight 353.39 g/mol.
- Intended for studies of fatty acid uptake, transport, and metabolic disease mechanisms.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.