Learn More
Cayman Chemical Fatty Acid ethyl ester Standa
SDP
Supplier:  Cayman Chemical 10008188100 MG
Encompass_Preferred
A pack that contains nine vials EAch containing 100 mg of an individual fatty acid ethyl ester; contains one vial EAch of stearic acid ethyl ester, myristic acid ethyl ester, linoleic acid ethyl ester, α-linolenic acid ethyl ester, arachidonic acid ethyl ester, oleic acid ethyl ester, palmitic acid ethyl ester, lauric acid ethyl ester, or palmitoleic acid ethyl ester
Catalog No. 50-288-9290