Learn More
Equitech Bio Inc  Fatty Acid Free Heat Shock Bovine Serum Albumin Powder, 50g
SDP
Supplier:  Equitech Bio Inc BAH660050
Encompass_Preferred
Heat Shocked Bovine Serum Albumin powdered uses a process of heat and pH adjustments to purify the product. If you don’t require a powder version, we also have 30% BSA solutions of the highest quality. 
Catalog No. NC9227912