Learn More
Medchemexpress LLC FAUC 346 | 474432-65-0 | 99.90% | 423.57 | 50 MG

Supplier: Medchemexpress LLC HY13880950MG
FAUC 346 is a highly selective D3 partial agonist with an EC50 of 1.5 nM, primarily intended for research use only.
- Highly selective D3 partial agonist
- D3-selective ligand with a Ki of 0.23 nM
- Solid appearance
- White to off-white color
- Store at 4°C, protected from light
- In solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light
- Ships at room temperature in continental US
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.