Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC FAUC 365 100mg | 474432-66-1 | 462.44 g·mol⁻¹ | C23H25Cl2N3OS | 100 MG
SDP

Supplier:  Medchemexpress LLC HY116020100MG

Encompass_Preferred

FAUC 365 is a research chemical described as a dopamine D3 receptor-selective antagonist used for in vitro pharmacology and receptor-binding studies. It is supplied in solid form and as a DMSO solution for biochemical assays.

  • Dopamine D3 receptor-selective antagonist.
  • High potency with reported Ki ≈ 0.5 nM at D3.
  • Purity 98.8%.
  • Chemical formula C23H25Cl2N3OS; molecular weight 462.44 g·mol⁻¹.
  • Available in small pack sizes, including 100 mg, and solution formats.
  • For research use only; not for human or clinical use.

Catalog No. 50-002-29887


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.