Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC FAUC 365 25mg | 474432-66-1 | 25 MG
SDP

Supplier:  Medchemexpress LLC HY11602025MG

Encompass_Preferred

FAUC 365 is a selective dopamine D3 receptor antagonist used as a research reagent to probe D3-mediated signaling in neuropharmacology and CNS disease models.

  • Highly selective dopamine D3 receptor antagonist (Ki = 0.5 nM).
  • Lower potency at related receptors (D4.4 Ki ≈ 340 nM; D2short Ki ≈ 2,600 nM; D2Long Ki ≈ 3,600 nM).
  • Chemical formula C23H25Cl2N3OS; molecular weight 462.44 g/mol.
  • Purity ~98.8% as supplied.
  • Appearance: off-white to light yellow solid.
  • Storage: powder -20°C for long-term storage; 4°C for shorter term.
  • Supplied as 25 mg research quantity.

Catalog No. 50-002-30049


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.