Learn More
Medchemexpress LLC HY-14768 50mg , Favipiravir CAS:259793-96-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1476850MG
Encompass_Preferred
HY-14768 50mg Favipiravir CAS: 259793-96-9 Favipiravir (T-705) is a potent viral RNA polymerase inhibitor, it is phosphoribosylated by cellular enzymes to its active form, Favipiravir-ribofuranosyl-5′-triphosphate (RTP). Favipiravir-RTP inhibits the influenza viral RNA-dependent RNA polymerase (RdRP) activity with an IC50 of 341 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0882