Learn More
Selleck Chemical LLC FB23-2 5mg
SDP
Supplier:  Selleck Chemical LLC S88375MG
Encompass_Preferred
FB23-2 is a potent and selective FTO inhibitor that directly binds to FTO and selectively inhibits FTO's N6-methyladenosine (m6A) demethylase activity with IC50 of 2.6 ?M. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3398