Learn More
Medchemexpress LLC HY-127103 5mg Medchemexpress, FB23-2 CAS:2243736-45-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1271035MG
Encompass_Preferred
Medchemexpress, HY-127103 5mg FB23-2 CAS:2243736-45-8 FB23-2 is a potent and selective inhibitor of mRNA N6-methyladenosine (m6A) demethylase FTO, with an IC50 of 2.6 μM. FB23-2 has anti-proliferation activity. FB23-2 can be used for the research of acute myeloid leukemia (AML). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1124