Learn More
TISSUE CULTURE BIOLOGICALS FBS 500 ML BOTTLE
Supplier:  TISSUE CULTURE BIOLOGICALS 101
Encompass
Fetal Bovine Serum (500ml) or FBS is aseptically collected by cardiac puncture closed system, in the United States. Tissue Culture Biologicals is one of the few serum companies that collect and process Fetal Bovine Blood directly at the abattoirs. All fetal bovine serum products are obtained in approved abattoirs from cattle that have been tested to be free from disease. All bovine serum lots are viral tested as per 9CFR 113-53. Serum is endotoxin, mycoplasma screened and selected for its ability to support growth of a wide range of cell culture by the harvest to plant ratio method. Hemoglobin content <15 mg/dL. Endotoxin level < 2 EU/m
Catalog No. NC0327704