Learn More
ATCC FBS, ES Cell Qualified
Catalog No. 502383967
Encompass
Fetal Bovine Serum, ES Cell Qualified is tested for its ability to support the growth and maintenance of embryonic stem cells in an undifferentiated state. Undifferentiated embryonic stem cells are determined by colony morphology and expression of three pluripotent markers. The results are compared to a control lot of serum. The serum is triple-filtered through 0.1 μM filters and each lot of fetal bovine serum is tested for sterility. Fetal bovine serum is manufactured from fetal bovine blood collected in USDA-inspected abattoirs located in the United States.
Catalog No. 50-238-3967 Supplier ATCC Supplier No. SCRR302020
Fetal Bovine Serum, ES Cell Qualified is tested for its ability to support the growth and maintenance of embryonic stem cells in an undifferentiated state. Undifferentiated embryonic stem cells are determined by colony morphology and expression of three pluripotent markers. The results are compared to a control lot of serum. The serum is triple-filtered through 0.1 μM filters and each lot of fetal bovine serum is tested for sterility. Fetal bovine serum is manufactured from fetal bovine blood collected in USDA-inspected abattoirs located in the United States.