Learn More
Norgen Biotek Corp FBS Exosome Depletion Kit I (Slurry Format), 6 Prep
Supplier:  Norgen Biotek Corp 61100
Encompass_Preferred
Norgen's FBS Exosome Depletion Kits (Slurry Format) constitute an all-in-one system for the depletion of cow's exosomes from FBS prior to using it as a growth supplement in your culture medium. The FBS recovered from the depletion process is exosome-depleted and does not contain any quantifiable bovine miRNAs. Moreover, the exosome-depleted FBS will support the growth of your cells of interest similar to the non-depleted FBS. Norgen's kits allows for the depletion of different FBS volumes with a maximum volume ranging from 140 mL to 280 mL. The depletion is based on Norgen’s proprietary resin. These kits do not require ultracentrifugation, special instrumentation, precipitation reagents or any protease treatments. More importantly, the depletion process is an inexpensive method for depletion of your own FBS, compared to the current ready-to-use exosome-depleted media available on the market.
Catalog No. NC0969852