Learn More
OriGene Fbx32 (FBXO32) Human siRNA Oligo Duplex (Locus ID 114907) 1 kit
SDP
Supplier:  OriGene SR314473
Encompass_Preferred
Components: FBXO32 (Human) - 3 unique 27mer siRNA duplexes - 2 nmol each (Locus ID 114907)Included - SR30004, Trilencer-27 Universal Scrambled Negative Control siRNA Duplex - 2 nmolIncluded - SR30005, RNAse free siRNA Duplex Resuspension Buffer - 2 ml
Catalog No. NC3233827