Learn More
Medchemexpress LLC Fc-11 (PROTAC FAK degrader) | 2271035-37-9 | ≥98.0% | 949.92 g/mol | C41H42F3N13O9S | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1393445MG
Encompass_Preferred
FC-11 is a PROTAC (proteolysis-targeting chimera) that induces targeted degradation of focal adhesion kinase (FAK). It links a CRBN-recruiting pomalidomide moiety to a FAK-binding ligand via a chemical linker to achieve potent cellular degradation (reported DC90 ≈ 1 nM), and is intended for biochemical and cellular studies of FAK biology.
  • Potent FAK degradation with DC90 ≈ 1 nM.
  • CRBN-mediated mode of action via pomalidomide ligand.
  • High purity suitable for biological assays (≥98.0%).
  • Molecular weight 949.92 g/mol and formula C41H42F3N13O9S.
  • Supplied in small research quantities, for example 5 MG packs.
Catalog No. 50-002-23075