Learn More
Medchemexpress LLC Fc-116 | 2417298-29-2 | MFCD20612185 | 98.0% | 369.39 g/mol | C21H20FNO4 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY15506810MG
Encompass_Preferred
FC-116 is a small-molecule tubulin inhibitor supplied for research use with demonstrated antitumor activity in preclinical models. It is provided as a high-purity solid (approx. 98.0%) and includes batch-specific analytical documentation for laboratory research. Handle according to institutional safety procedures; not for human use.
  • Acts as a tubulin inhibitor with reported antitumor activity.
  • High analytical purity (98.0%).
  • Supplied as an off-white to light yellow solid.
  • Suitable for in vitro and preclinical research applications.
  • Provided with a certificate of analysis for batch-specific data.
  • Identified by CAS number 2417298-29-2 for unambiguous reference.
Catalog No. 50-002-87360