Learn More
BD Cell Analysis 3P FC BEADS 7-COLOR KIT
Supplier:  BD Cell Analysis 3P 662961
Encompass
BD FC Beads are fluorescent beads thatenable the software to calculate afluorescence compensation matrix duringsetup of the flow cytometer
Catalog No. NC1922969