Learn More
ACROBiosystems CHO/Human CD32a (131H) Stable Cell Line (Medium Expression), 2 vials (≥5x10^6 cells/vial)
Supplier:  ACROBiosystems SCCHOATP061M2VIALS
Encompass_Preferred
A CHO stable cell line engineered to express full-length human Fc gamma RIIa (CD32a, 131H) at medium expression. Supplied frozen in serum-free cryopreservation medium as two vials, each containing at least 5x10^6 cells. Intended for cell-based CD32a (131H) binding assays, CD32a-mediated crosslinking studies, and testing agonist antibodies for CD32a-dependent enhancement of activity.
  • Supplied as 2 vials, 1 mL cryopreserved cells (≥5x10^6 cells per vial).
  • Frozen storage in liquid nitrogen.
  • CHO stable cell line expressing human CD32a (131H) at medium level.
  • Adherent growth properties.
  • Hygromycin B selection (20 μg/mL).
  • Complete growth medium: F-12K with 10% FBS.
  • Mycoplasma- and sterility-tested; negative results.
  • For research use only; commercial use requires separate license.
Catalog No. 50-235-7340