Learn More
Genscript Corporation Fc gamma RIIIA/CD16a F176Bi
Supplier:  Genscript Corporation Z04082100
Encompass
Human Fc gamma RIIIA/CD16a Protein is a receptor for the Fc region of IgG Binds complexed or aggregated IgG and also monomeric IgG Mediates antibody-dependent cellular cytotoxicity (ADCC) and other antibody-dependent responses such as phagocytosis
Catalog No. 50-000-58475