Learn More
Cusabio Technology LLC Human Fc region of immunoglobulin G,Fcγ ELISA Kit
Supplier:  Cusabio Technology LLC CSBE11223H
Encompass_Preferred
This Human Fcγ ELISA Kit was designed for the quantitative measurement of Human Fcγ protein in serum, plasma. It is a Competitive ELISA kit, its detection range is 0.027 μg/mL-20 μg/mL and the sensitivity is 0.024 μg/mL. One kit contains 96 wells.
Catalog No. NC2952859