Learn More
Selleck Chemical LLC FCCP 10mg 370-86-5 Trifluoromethoxy carbonylcyanide phenylhydrazone
SDP
Supplier:  Selleck Chemical LLC S827610MG
Encompass_Preferred
FCCP (Trifluoromethoxy carbonylcyanide phenylhydrazone) is a potent uncoupler of oxidative phosphorylation in mitochondria that disrupts ATP synthesis by transporting protons across cell membranes. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5882