Learn More
Enzo Life Sciences FCCP, (10mg), CAS Number: 370-86-5
SDP
Supplier:  Enzo Life Sciences BMLCM1200010
Encompass_Preferred
MPTP inhibitor. A potent reversible inhibitor of mitochondrial oxidative phosphorylation. FCCP is a useful tool for depolarizing mitochondrial membrane potential. Treatment of cells with FCCP at varying concentrations leads to partial (100 nM) or complete (10 µM) depolarization and apoptosis. Alternative Name: Carbonylcyanide-4-(trifluoromethoxy)-phenylhydrazone. Formula: C10H5F3N4O. MW: 254.2. CAS Number: 370-86-5. Purity: ≥98% (HPLC). Appearance: Yellow solid. Solubility: Soluble in DMSO. Long Term Storage: Ambient
Catalog No. 50-103-1773