Learn More
ACROBiosystems ACROBiosystems Human Fc gamma RIIIB / CD16b (NA1) Protein, His Tag (SPR & BLI verified)
Supplier: ACROBiosystems CDBH522750UG
Recombinant Protein;50UG;Human CD16b (NA1), His Tag (SPR & BLI verified) (CDB-H5227) is expressed from human HEK293 cells. It contains AA Gly 17 - Ser 200 (Accession # AAA35881.1).The NA1 form of the CD16b differ with the NA2 form of CD16b in AA36, 65, 82, and 106. The NA1 form carries R36, N65, D82, and V106, while the NA2 form carries S36, S65, N82, and I106.;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 22.8 kDa. The protein migrates as 35-45 KDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD16b (NA1), His Tag (SPR & BLI verified) (Cat. No. CDB-H5227) on CM5 Chip via anti-His antibody, can bind Rituximab with an affinity constant of 8.17 μM as determined in a SPR assay (Biacore T200) (QC tested).
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.