Learn More
Medchemexpress LLC FCGRT-B2M Heterodime 1mg | 1mg
SDP
Supplier:  Medchemexpress LLC HYP769201MG
Encompass_Preferred
The FCGRT protein delivers passive immunity by binding to monomeric IgG and promoting its absorption from milk It forms an FcRn-IgG complex which is transcytosis by the intestinal epithelium and releases IgG into the bloodstream or interstitial fluid FCGRT-B2M Heterodimer Protein Cynomolgus (HEK293 His) is a recombinant protein dimer complex containing cynomolgus-derived FCGRT-B2M Heterodimer protein expressed by HEK293 with C-His labeled tag FCGRT-B2M Heterodimer Protein Cynomolgus (HEK293 His) has molecular weight of 35 12 kDa respectively
Catalog No. 50-001-86165