Learn More
Med Vet International Miltex Meisterhand Kantrowitz Thoracic Hemostatic Forceps, 7.5in., Delicate Right Angle Jaws
Supplier:  Med Vet International MILMH25836
Encompass_Preferred
The Miltex MeisterHand MH25-836 Kantrowitz thoracic forceps are made of OR grade German stainless steel and measure 7.5" in length. They have a finger ring handle with a ratchet lock and delicate right angle jaws.
Catalog No. 50-283-1403